missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human INSL3 (NP_005534.2) VersaClone cDNA

Product Code. 18710297
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18710297 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18710297 Supplier R&D Systems™ Supplier No. RDC3271

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Gene INSL3
Accession Number NP_005534.2
Gene ID (Entrez) 3640
Gene Alias insulin-like 3, insulin-like 3 (Leydig cell), Ley I-L, leydig insulin -like hormone, leydig insulin -like peptide, Leydig insulin-like peptide, Ley-I-L, MGC119818, MGC119819, Relaxin-like factor, relaxin-like factor b, RLF, RLFley-I-L, RLNL
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.