Learn More
Human IVNS1ABP (Influenza Virus NS1A Binding Protein) knockout HeLa cell lysate, 1 Kit
Product Code. 32147714 Shop All Abcam Products
Product Code. 32147714 Supplier Abcam Supplier No. ab2584781Kit
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Human IVNS1ABP (Influenza Virus NS1A Binding Protein) knockout HeLa cell/tissue lysate.
  • Species: Human
  • Tissue: IVNS1ABP (Influenza Virus NS1A Binding Protein) knockout HeLa