missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human JAM-C Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB11891R-100UG
This item is not returnable.
View return policy
Description
JAM-C Monoclonal specifically detects JAM-C in Human samples. It is validated for Flow Cytometry.
Specifications
| JAM-C | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| CD323, JAM-2, JAM3, JAMC, JAM-CFLJ14529, junctional adhesion molecule 3JAM-3, junctional adhesion molecule C | |
| Mouse myeloma cell line NS0-derived recombinant human JAM-C Val32-Asn241(Ala149Pro) Accession # Q9BX67 | |
| 100 μg | |
| 83700 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 208212 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q9BX67 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human JAM-C in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction