Learn More
Novus Biologicals™ Human Lung Tissue MicroArray (Cancer)
Product Code. 18746163 Shop All Bio Techne Products
Change view
Click to view available options
1 Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18746163 1 Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18746163 Supplier Novus Biologicals™ Supplier No. NBP230277
  • ONLINE EXCLUSIVE PRICE
    342.00€
    323.40€ / Pack of 1
    Save 18.60€
    5% Off
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
Applications: Immunohistochemistry-Paraffin
TRUSTED_SUSTAINABILITY
Specifications
Description 59 Carcinoma, adenocarcinoma, carcinosarcoma
Quantity 1 Slide
Species Human
Tissue Condition Cancer
Type Lung
For Use With (Application) Immunohistochemistry-Paraffin
Dimensions (LxW) 2.0 mm