missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Ly6K Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6648-SP
This item is not returnable.
View return policy
Description
Ly6K Monoclonal specifically detects Ly6K in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| Ly6K | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q17RY6 | |
| LY6K | |
| Chinese hamster ovary cell line CHO-derived recombinant human Ly6K Met1-Gly138 Accession # Q17RY6 | |
| 25 μg | |
| Primary | |
| Detects human Ly6K in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 750747 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cancer/testis antigen 97, CO16, CT97, FLJ35226, HSJ001348, Ly-6K, lymphocyte antigen 6 complex, locus K, lymphocyte antigen 6K | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 54742 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction