missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MCPH1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF3998-SP
This item is not returnable.
View return policy
Description
MCPH1 Polyclonal specifically detects MCPH1 in Human samples. It is validated for Western Blot.
Specifications
| MCPH1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BRCT-repeat inhibitor of TERT expression 1, BRIT1, FLJ12847, MCT, microcephalin, microcephalin 1, microcephaly, primary autosomal recessive 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 79648 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q8NEM0 | |
| MCPH1 | |
| E. coli-derived recombinant human MCPH1 Met1-Lys250 Accession # Q8NEM0 | |
| 25 μg | |
| Primary | |
| Detects human, mouse and rat MCPH1 in Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction