missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human MDM2/HDM2 (AAM78554) VersaClone cDNA

Product Code. 18718679
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18718679 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18718679 Supplier R&D Systems™ Supplier No. RDC2746
  • ONLINE EXCLUSIVE PRICE
    328.00€
    309.75€ / 10µg
    Save 18.25€
    5% Off
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

  • Also known as: HDMX; hdm2; ACTFS
  • MDM2 is a nuclear-localized E3 ubiquitin ligase.
  • It can promote tumor formation by targeting tumor suppressor proteins, such as p53, for proteasomal degradation.
  • Overexpression or amplification of this locus is detected in a variety of different cancers.
  • Alternatively spliced transcripts encoding different proteins have been described.
  • The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
  • For reasearch use only. Not for use in humans.
TRUSTED_SUSTAINABILITY

Specifications

Gene MDM2/HDM2
Accession Number AAM78554
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 1489 bp
Gene ID (Entrez) 4193
Gene Alias ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.