missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MMP1 native Protein from human fibroblasts

Product Code. 16068076
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16068076 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16068076 Supplier Abnova™ Supplier No. P4919.10ug

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Used for Func, SDS-PAGE

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. This gene encodes a secreted enzyme which breaks down the interstitial collagens, types I, II, and III. The gene is part of a cluster of MMP genes which localize to chromosome 11q22.3. Alternative splicing results in multiple transcript variants.[provided by RefSeq]

Specifications

Concentration 0.2 mg/mL
For Use With (Application) Functional Study, SDS-PAGE
Formulation In 50mM Tris-HCl, 300mM NaCl, 5mM CaCl
Gene ID (Entrez) 4312
Molecular Weight (g/mol) 52-56kDa
Name MMP1 (Human) Native Protein
Preparation Method Native protein purified from human fibroblasts
Purification Method Affinity Chromatography/Ion Exchange Chromatography/Gelfiltration
Quality Control Testing 15% SDS-PAGE Stained with Coomassie Blue
Quantity 10 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CLG/CLGN
Common Name MMP1
Gene Symbol MMP1
Biological Activity ≥ 100mU/mg (upon trypsin activation)
Species Human
Protein Tag None
Expression System Native protein purified from human fibroblasts
Form Liquid
Purity or Quality Grade >90% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.