missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Peroxiredoxin 5 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3774-SP
This item is not returnable.
View return policy
Description
Peroxiredoxin 5 Polyclonal specifically detects Peroxiredoxin 5 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Peroxiredoxin 5 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ACR1, ACR1Peroxisomal antioxidant enzyme, Alu corepressor 1, Alu co-repressor 1, Antioxidant enzyme B166, AOEB166, AOEB166Peroxiredoxin V, AOPP, B166, EC 1.11.1.15, Liver tissue 2D-page spot 71B, MGC117264, MGC142283, MGC142285, peroxiredoxin 5, peroxiredoxin-5, mitochondrial, PLPThioredoxin peroxidase PMP20, PMP20, PRDX5, PRDX6, prx-V, PRXV, SBBI10, Thioredoxin reductase, TPx type VI | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 25824 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| P30044 | |
| PRDX5 | |
| E. coli-derived recombinant human Peroxiredoxin 5 Met1-Leu214 Accession # P30044 | |
| 25 μg | |
| Primary | |
| Detects endogenous human and mouse Peroxiredoxin 5 in Western blots. In Western blots, this antibody shows no cross-reactivity with recombinant human Peroxiredoxin 1, 2, 3, or 6. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction