Learn More
Human/Mouse RASSF2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF5639-SP
This item is not returnable.
View return policy
Description
RASSF2 Polyclonal specifically detects RASSF2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| RASSF2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| KIAA0168DKFZp781O1747, Ras association (RalGDS/AF-6) domain family 2, Ras association (RalGDS/AF-6) domain family member 2, ras association domain-containing protein 2, RASFADIN | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9770 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P50749 | |
| RASSF2 | |
| E. coli-derived recombinant human RASSF2 Lys100-Gly230 Accession # P50749 | |
| 25 μg | |
| Primary | |
| Detects endogenous human and mouse RASSF2 in Western blots. | |
| Human, Mouse | |
| IgG |