missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat BNIP3L Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF4030
This item is not returnable.
View return policy
Description
BNIP3L Polyclonal specifically detects BNIP3L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| BNIP3L | |
| Polyclonal | |
| Unconjugated | |
| O60238 | |
| BNIP3L | |
| E. coli-derived recombinant human BNIP3L Ser2-Glu184 Accession # O60238 | |
| 100 μg | |
| Primary | |
| Detects endogenous human, mouse and rat BNIP3L in Western blots. In Western blots, less than 10% cross-reactivity to recombinant human BNIP3L is observed. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Adenovirus E1B19K-binding protein B5, BCL2/adenovirus E1B 19 kDa protein-interacting protein 3A, BCL2/adenovirus E1B 19 kDa protein-interacting protein 3-like, BCL2/adenovirus E1B 19-kd protein-interacting protein 3a, BCL2/adenovirus E1B 19kDa interacting protein 3-like, BNIP3a, BNIP3H, Nip3L, NIP3-like protein X, Nix, NIXBCL2/adenovirus E1B 19kD-interacting protein 3-like | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 665 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction