missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat DGCR2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
199.00€ - 440.00€
Specifications
| Antigen | DGCR2 |
|---|---|
| Clone | 1019908 |
| Dilution | Western Blot 2 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18702967
![]() |
R&D Systems
MAB10443-SP |
25 μg |
199.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18742047
![]() |
R&D Systems
MAB10443-100 |
100 μg |
440.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DGCR2 Monoclonal specifically detects DGCR2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| DGCR2 | |
| Western Blot 2 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| DGS-C, DiGeorge syndrome critical region gene 2, IDDDKFZp686I1730, integral membrane protein deleted in DiGeorge syndrome, integral membrane protein DGCR2/IDD, KIAA0163, KIAA0163DiGeorge syndrome critical region protein 2, LAN, SEZ-12 | |
| Chinese Hamster Ovary cell line CHO-derived human DGCR2, Glu22-Ala349, Accession # P98153 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1019908 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| P98153 | |
| 9993 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title