missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL20/MIP-3 alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 12 publications
Brand: R&D Systems MAB360-100
This item is not returnable.
View return policy
Description
CCL20/MIP-3 alpha Monoclonal specifically detects CCL20/MIP-3 alpha in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| CCL20/MIP-3 alpha | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARC, LARCLiver and activation-regulated chemokine, MIP3 alpha, MIP-3 alpha, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, SCYA20Small-inducible cytokine A20, small inducible cytokine subfamily A (Cys-Cys), member 20, ST38 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 6364 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Neutralization, ELISA | |
| 67310 | |
| Western Blot 1 ug/mL, Neutralization 0.2-2 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P78556.1 | |
| CCL20 | |
| E. coli-derived recombinant human CCL20/MIP-3 alpha Ala27-Met96 (Asn95Asp) Accession # P78556.1 | |
| 100 μg | |
| Primary | |
| Detects human CCL20/MIP-3 alpha in ELISAs and Western blots. In ELISAs, no cross-reactivity with recombinant human (rh) CCL19, rhCXCL1, rhCCL16, rhCCL11, recombinant mouse CCL20/MIP-3 alpha , or recombinant rat CCL20/MIP-3 alpha is observed. | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction