missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat GATE-16 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB9035
This item is not returnable.
View return policy
Description
GATE-16/GABARAPL2 Monoclonal specifically detects GATE-16/GABARAPL2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| GATE-16/GABARAPL2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Apg8p2, ATG8, ATG8C, FLC3A, GABARAPL2, gamma-aminobutyric acid receptor-associated protein-like 2, Ganglioside expression factor 2, GATE16, GATE-16, GEF2, GEF-2, GEF2GEF-2, General Protein Transport Factor P16, Golgi-Associated ATPase Enhancer Of 16 KDa, MAP1 light chain 3 related protein, MAP1 light chain 3-related protein | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11345 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 853746 | |
| Western Blot 2 ug/mL | |
| P60520 | |
| GABARAPL2 | |
| E.coli-derived recombinant human GATE16 Accession # P60520 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat GATE-16 in Western blot. | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction