missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat GRK2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4339-SP
This item is not returnable.
View return policy
Description
GRK2 Polyclonal specifically detects GRK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Knockout Validated.
Specifications
| GRK2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ADRBK1, adrenergic, beta, receptor kinase 1, BARK, BARK1, BARK1beta-ARK-1, Beta-ARK-1, BETA-ARK1, EC 2.7.11, EC 2.7.11.15, FLJ16718, G-protein coupled receptor kinase 2, GRK2beta-adrenergic receptor kinase 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 156 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunoassay | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Knockout Validated | |
| P25098 | |
| ADRBK1 | |
| E. coli-derived recombinant human GRK2 Pro468-Leu689 Accession # P25098 | |
| 25 μg | |
| Primary | |
| Detects endogenous human, mouse, and rat GRK2. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction