missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat IKK alpha Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF3768-SP
This item is not returnable.
View return policy
Description
IKK alpha Polyclonal specifically detects IKK alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| IKK alpha | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CHUK, conserved helix-loop-helix ubiquitous kinaseIKBKA, EC 2.7.11, EC 2.7.11.10, IkappaB kinase, I-kappa-B kinase 1, I-kappa-B kinase alpha, IkBKA, IkBKalpha, IKK1, IKK1I-kappa-B kinase-alpha, IKK-A, IKKAIkB kinase alpha subunit, IKK-alphainhibitor of nuclear factor kappa-B kinase subunit alpha, NFKBIKA, NFKBIKAIKK-a kinase, Nuclear factor NFkappaB inhibitor kinase alpha, Nuclear factor NF-kappa-B inhibitor kinase alpha, TCF16, TCF-16, Transcription factor 16 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1147 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| O15111 | |
| CHUK | |
| E. coli-derived recombinant human IKK alpha Gly375-Glu745 Accession # O15111 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat IKK alpha . Does not cross-react with recombinant human (rh) IKK beta , rhIKK gamma , or rhIKK epsilon . | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction