missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Lipocalin-2/NGAL Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 14 publications
Brand: R&D Systems AF1757
This item is not returnable.
View return policy
Description
Lipocalin-2/NGAL Polyclonal specifically detects Lipocalin-2/NGAL in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| Lipocalin-2/NGAL | |
| Polyclonal | |
| Unconjugated | |
| P80188 | |
| LCN2 | |
| Mouse myeloma cell line NS0-derived Recombinant Human Lipocalin-2/NGAL, Gln21-Gly198, Accession # P80188 | |
| 100 μg | |
| Primary | |
| Detects human Lipocalin-2/NGAL in direct ELISAs and Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Simple Western 2 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| 24p3, 25 kDa alpha-2-microglobulin-related subunit of MMP-9, HNL, LCN2, lipocalin 2, lipocalin 2 (oncogene 24p3), Lipocalin2, Lipocalin-2, migration-stimulating factor inhibitor, MSFI, neutrophil gelatinase-associated lipocalin, NGAL, NGALlipocalin-2, Oncogene 24p3, p25, Siderocalin, Uterocalin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3934 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction