missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat p27/Kip1 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC2256S-100UG
This item is not returnable.
View return policy
Description
p27/Kip1 Monoclonal specifically detects p27/Kip1 in Human, Mouse, Rat samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| p27/Kip1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P46527 | |
| CDKN1B | |
| E. coli-derived recombinant human p27 Met1-Thr198 Accession # P46527 | |
| RUO | |
| 1027 | |
| Human, Mouse, Rat | |
| IgG2b |
| Flow Cytometry | |
| 225501 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CDKN1B, CDKN4, cyclin-dependent kinase inhibitor 1B, cyclin-dependent kinase inhibitor 1B (p27, Kip1), Cyclin-dependent kinase inhibitor p27, Kip1, KIP1P27KIP1, MEN1B, MEN4, p27Kip1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat p27 in Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction