missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Tmp21/p23 Antibody, R&D Systems™

Mouse Monoclonal Antibody
170.00€ - 398.00€
Specifications
| Antigen | Tmp21/p23 |
|---|---|
| Clone | 805125 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18773609
![]() |
R&D Systems
MAB7630-SP |
25 μg |
170.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18704899
![]() |
R&D Systems
MAB7630-100 |
100 μg |
398.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tmp21/p23 Monoclonal specifically detects Tmp21/p23 in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.Specifications
| Tmp21/p23 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human, Mouse, Rat | |
| P49755 | |
| 10972 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 805125 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 21 kDa transmembrane-trafficking protein, p23, P24(DELTA), p24delta, S31I125, S31III125, TMED10, TMP21Tmp-21-I, transmembrane emp24 domain-containing protein 10, transmembrane emp24-like trafficking protein 10 (yeast), Transmembrane protein Tmp21,21 kDa transmembrane trafficking protein | |
| E.coli-derived recombinant human Tmp21/p23 Ile32-Arg185 Accession # P49755 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Tmp21/p23 in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title