missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse TL1A/TNFSF15 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
208.00€ - 541.00€
Specifications
| Antigen | TL1A/TNFSF15 |
|---|---|
| Clone | 293327 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Neutralization 1 ug/mL |
| Applications | Western Blot, Neutralization |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18700928
![]() |
R&D Systems
MAB7441-SP |
25 μg |
208.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18130461
|
R&D Systems
MAB7441 |
100 μg |
541.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TL1A/TNFSF15 Monoclonal specifically detects TL1A/TNFSF15 in Human, Mouse samples. It is validated for Western Blot, Neutralization.Specifications
| TL1A/TNFSF15 | |
| LYOPH | |
| Western Blot, Neutralization | |
| Unconjugated | |
| Rat | |
| Human, Mouse | |
| NP_796345.3 | |
| 9966 | |
| E. coli-derived recombinant mouse TL1A/TNFSF15 Ile94-Leu270 Accession # NP_796345.3 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse TL1A/TNFSF15 in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant human TL1A. |
| 293327 | |
| Western Blot 1 ug/mL, Neutralization 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, TNFSF15, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI, VEGI192A, VEGITNF ligand-related molecule 1 | |
| TNFSF15 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title