missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ human MuRF1/TRIM63 (NP_115977.2) VersaClone cDNA
Specifications
Specifications
| Gene | MuRF1/TRIM63 |
| Accession Number | NP_115977.2 |
| Gene ID (Entrez) | 84676 |
| Gene Alias | EC 6.3.2.-, IRFMURF2, Iris RING finger protein, MuRF1, MuRF-1, muscle specific ring finger protein 2, Muscle-specific RING finger protein 1, RING finger protein 28 FLJ32380, RNF28E3 ubiquitin-protein ligase TRIM63, SMRZMURF-1, Striated muscle RING zinc finger protein, tripartite motif containing 63, tripartite motif-containing 63, Tripartite motif-containing protein 63 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?