missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat CXCR3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8109
This item is not returnable.
View return policy
Description
CXCR3 Monoclonal specifically detects CXCR3 in Rat samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| CXCR3 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| CD183, CD183 antigen, chemokine (C-X-C motif) receptor 3, chemokine (C-X-C) receptor 3, CKR-L2IP-10 receptor, CMKAR3, C-X-C chemokine receptor type 3, CXCR3, CXC-R3, CXCR-3, G protein-coupled receptor 9CD182, GPR9, GPR9Mig-R, Interferon-inducible protein 10 receptor, IP10 receptor, IP10-R, Mig receptor, MigR | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects rat CXCR3 in ELISA. Stains rat CXCR3 transfected cells but not irrelevant transfectants by Flow Cytometry. | |
| Rat | |
| Purified |
| Flow Cytometry, CyTOF | |
| 868013 | |
| Unconjugated | |
| Q9J119 | |
| CXCR3 | |
| NS0 mouse myeloma cell line transfected with rat CXCR3 Accession # Q9J119 | |
| 100 μg | |
| 2833 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction