Learn More
R&D Systems™ Human Neuroligin 2/NLGN2 (NP_065846.1) VersaClone cDNA
Product Code. 18791557
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18791557 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18791557 Supplier R&D Systems™ Supplier No. RDC3674
Specifications
Gene Neuroligin 2/NLGN2
Accession Number NP_065846.1
Gene ID (Entrez) 57555
Gene Alias EC 3.1.1, EC 3.1.1.1, KIAA1366neuroligin-2, neuroligin 2
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg