Learn More
R&D Systems™ Human Neuroligin 2/NLGN2 (NP_065846.1) VersaClone cDNA
Specifications
Specifications
| Gene | Neuroligin 2/NLGN2 |
| Accession Number | NP_065846.1 |
| Gene ID (Entrez) | 57555 |
| Gene Alias | EC 3.1.1, EC 3.1.1.1, KIAA1366neuroligin-2, neuroligin 2 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 10 μg |