missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NGL-1/LRRC4C Antibody, R&D Systems™

Sheep Polyclonal Antibody
201.00€ - 511.00€
Specifications
| Antigen | NGL-1/LRRC4C |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18748457
![]() |
R&D Systems
AF4899-SP |
25 μg |
201.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18071892
![]() |
R&D Systems
AF4899 |
100 μg |
511.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NGL-1/LRRC4C Polyclonal specifically detects NGL-1/LRRC4C in Human samples. It is validated for Western Blot.Specifications
| NGL-1/LRRC4C | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Sheep | |
| Human | |
| KIAA1580NGL-1NGL1Netrin-G1 ligand, leucine rich repeat containing 4C, leucine-rich repeat-containing protein 4C, LRRC4C, NGL1 | |
| LRRC4C | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9HCJ2 | |
| 57689 | |
| Mouse myeloma cell line NS0-derived recombinant human NGL-1/LRRC4C Gln45-Lys527, predicted Accession # Q9HCJ2 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human NGL-1/LRRC4C in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title