missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NKCC1/SLC12A2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8528
This item is not returnable.
View return policy
Description
NKCC1/SLC12A2 Monoclonal specifically detects NKCC1/SLC12A2 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| NKCC1/SLC12A2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Basolateral Na-K-Cl symporter, BSC, BSC2, Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1, NKCC1MGC104233, SLC12A2, solute carrier family 12 (sodium/potassium/chloride transporters), member 2, solute carrier family 12 member 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6558 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 915911 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P55011 | |
| SLC12A2 | |
| NS0 mouse myeloma cell line transfected with human NKCC1/SLC12A2 Met1-Ser1212 Accession # P55011 | |
| 100 μg | |
| Primary | |
| Stains NKCC1/SLC12A2 transfectants but not irrelevant transfectants in flow cytometry. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction