missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human NLRP2/NALP2 (NP_060322.1) VersaClone cDNA

Product Code. 18774477
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18774477 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18774477 Supplier R&D Systems™ Supplier No. RDC3758
  • ONLINE EXCLUSIVE PRICE
    853.00€
    806.40€ / 10µg
    Save 46.60€
    5% Off
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Gene NLRP2/NALP2
Accession Number NP_060322.1
Gene ID (Entrez) 55655
Gene Alias CLR19.9, FLJ20510, NACHT, leucine rich repeat and PYD containing 2, NACHT, LRR and PYD domains-containing protein 2, NALP2, NALP2NACHT, LRR and PYD containing protein 2, NBS1, NBS1PYRIN-containing APAF1-like protein 2, NLR family, pyrin domain containing 2, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 2, PAN1, PAN1Nucleotide-binding site protein 1, PYPAF2, PYPAF2PYRIN domain and NACHT domain-containing protein 1
Content And Storage Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.