missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NLRP6/NALP6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
184.00€ - 378.00€
Specifications
| Antigen | NALP6 |
|---|---|
| Clone | 920631 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18788408
![]() |
R&D Systems
MAB9145-SP |
25 μg |
184.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18708339
![]() |
R&D Systems
MAB9145 |
100 μg |
378.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NALP6 Monoclonal specifically detects NALP6 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| NALP6 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Angiotensin II/Vasopressin Receptor, AVR, CLR11.4, NACHT, Leucine Rich Repeat And PYD Containing 6, NACHT, LRR And PYD Containing Protein 6, NACHT, LRR And PYD Domains-Containing Protein 6, NALP6, NAVR, NAVR/AVR, NLR Family, Pyrin Domain Containing 6, NLRP6, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 6, PAN3, PYPAF5, PYRIN-Containing APAF1-Like Protein 5 | |
| NLRP6 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 920631 | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| P59044 | |
| 171389 | |
| E.coli-derived recombinant human NLRP6/NALP6 Lys35-Arg193 Accession # P59044 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human NLRP6/NALP6 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title