missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human NLRP7 (NP_996611.2) VersaClone cDNA

Product Code. 18731787
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18731787 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18731787 Supplier R&D Systems™ Supplier No. RDC3609

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Gene NLRP7
Accession Number NP_996611.2
Gene ID (Entrez) 199713
Gene Alias CLR19.4, MGC126471, NACHT, leucine rich repeat and PYD containing 7, NACHT, LRR and PYD containing protein 7, NACHT, LRR and PYD domains-containing protein 7, NALP7, NALP7MGC126470, NLR family, pyrin domain containing 7, NOD12, NOD12HYDM, Nucleotide-binding oligomerization domain protein 12, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 7, PAN7, PYPAF3, PYPAF3FLJ94610, PYRIN-containing APAF1-like protein 3
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.