Learn More
Human NOD1 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB7090-SP
Product Code. 18721678
  • 214.00€ / 25µg
Description
NOD1 Monoclonal specifically detects NOD1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NOD1
Monoclonal
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Q9Y239
NOD1
E. coli-derived recombinant human NOD1 Lys830-Phe953 Accession # Q9Y239
25 μg
Primary
Detects human NOD1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human NOD2 is observed.
Human
Purified
Western Blot, Immunohistochemistry
626919
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CARD4, CARD4NLR family, CARD domain containing 1, caspase recruitment domain family, member 4, Caspase recruitment domain-containing protein 4, CLR7.1, NLRC1, nucleotide-binding oligomerization domain containing 1, nucleotide-binding oligomerization domain, leucine rich repeat and CARD domaincontaining 1, nucleotide-binding oligomerization domain-containing protein 1
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
10392
Sterile PBS to a final concentration of 0.5 mg/mL.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b