Learn More
Human NOSIP knockout HEK-293T cell lysate, 1 Kit
Product Code. 32154907 Shop All Abcam Products
Product Code. 32154907 Supplier Abcam Supplier No. ab2589991Kit
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Human NOSIP knockout HEK-293T cell/tissue lysate.
  • Species: Human
  • Tissue: NOSIP knockout HEK-293T