missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human NPSR1 (NP_997055) VersaClone cDNA
Description
- Also known as: GPRA; NPSR; VRR1; ASRT2; PGR14; GPR154
- NPSR1 is a member of the vasopressin/oxytocin subfamily of G protein-coupled receptors.
- It acts as a receptor for neuropeptide S and affects a variety of cellular processes through its signaling.
- Increased expression of NPSR1 in ciliated cells of the respiratory epithelium and in bronchial smooth muscle cells is associated with asthma.
- Polymorphisms in NPSR1 have also been associated with asthma susceptibility, panic disorders, inflammatory bowel disease, and rheumatoid arthritis.
- Alternatively spliced transcripts encoding different proteins have been described.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hNPSR1 |
| Accession Number | NP_997055 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1129 bp |
| Gene ID (Entrez) | 387129 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?