Learn More
R&D Systems™ Human OGDH (NP_002532.2) VersaClone cDNA
Specifications
Specifications
| Gene | OGDH |
| Accession Number | NP_002532.2 |
| Gene ID (Entrez) | 4967 |
| Gene Alias | 2-oxoglutarate dehydrogenase complex component E1, 2-oxoglutarate dehydrogenase, mitochondrial, AKGDH, alpha KGD, alpha-KGD, E1k, KGD1, OGDC, OGDC-E1, OGDH2, oxoglutarate (alpha-ketoglutarate) dehydrogenase (lipoamide), oxoglutarate decarboxylase, oxoglutarate dehydrogenase (succinyl-transferring) |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |