missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human OPA1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB11631-SP
This item is not returnable.
View return policy
Description
OPA1 Monoclonal antibody specifically detects OPA1 in Human samples. It is validated for Western Blot,Simple Western,Immunohistochemistry
Specifications
| OPA1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| BERHS, EC 3.6.5.5, FLJ12460, KIAA0567dynamin-like 120 kDa protein, mitochondrial, LargeG, lilr3, MGM1, mitochondrial dynamin-like GTPase, MTDPS14, NPG, NPGlargeG, NTG, NTGmitochondrial dynamin-like 120 kDa protein, optic atrophy 1 (autosomal dominant), Optic atrophy protein 1 | |
| Synthetic Peptide, Accession # O60313 | |
| 25 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Western Blot, Simple Western, Immunohistochemistry | |
| 1088319 | |
| Western Blot 2 μg/mL, Simple Western 100 μg/mL, Immunohistochemistry 3-25 μg/mL | |
| O60313 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 4976 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction