missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Osteopontin/OPN Antibody, R&D Systems™

Rabbit Monoclonal Antibody
211.00€ - 497.00€
Specifications
| Antigen | Osteopontin/OPN |
|---|---|
| Clone | 2579A |
| Dilution | Immunohistochemistry 3-25 ug/mL, Neutralization 0.5-1.5 ug/mL, Immunocytochemistry 3-25 ug/mL |
| Applications | Immunohistochemistry, Neutralization, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18702957
![]() |
R&D Systems
MAB14334-SP |
25 μg |
211.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18742137
![]() |
R&D Systems
MAB14334-100 |
100 μg |
497.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Osteopontin/OPN Monoclonal specifically detects Osteopontin/OPN in Human samples. It is validated for Immunohistochemistry, Neutralization, Immunocytochemistry.Specifications
| Osteopontin/OPN | |
| Immunohistochemistry 3-25 ug/mL, Neutralization 0.5-1.5 ug/mL, Immunocytochemistry 3-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| BNSP, Bone sialoprotein 1, Eta-1, MGC110940, Nephropontin, OPN, osteopontin, secreted phosphoprotein 1bone sialoprotein I, early T-lymphocyteactivation 1), secreted phosphoprotein-1 (osteopontin, bone sialoprotein), Spp1, SPP-1, SPP1/CALPHA1 fusion, Urinary stone protein, uropontin | |
| Synthetic peptide containing human Osteopontin/OPN, Accession # NP_000573.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2579A | |
| Immunohistochemistry, Neutralization, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| NP_000573.1 | |
| 6696 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title