missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human PACE4/PCSK6 (NP_002561) VersaClone cDNA

Product Code. 18782118
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18782118 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18782118 Supplier R&D Systems™ Supplier No. RDC2520

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.

  • The Human PCSK6 (NP_002561) VersaClone cDNA clone contains the complete ORF for PACE4, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The PACE4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
TRUSTED_SUSTAINABILITY

Specifications

Gene PACE4/PCSK6
Accession Number NP_002561
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Gene ID (Entrez) 5046
Gene Alias EC 3.4.21, EC 3.4.21.61, EC 3.4.21.75, PACE4, PACE4EC 3.4.21.-, Paired basic amino acid cleaving enzyme 4, paired basic amino acid cleaving system 4, PCSK6, Proprotein Convertase 6, proprotein convertase subtilisin/kexin type 6, SPC4, SPC4subtilisin-like protease, Subtilisin/kexin-like protease PACE4, Subtilisin-like proprotein convertase 4
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.