missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PD-L1/B7-H1 (Research Grade Atezolizumab Biosimilar) Antibody, R&D Systems™

Human Monoclonal Antibody
Brand: R&D Systems MAB10348-100
This item is not returnable.
View return policy
Description
PD-L1 Monoclonal specifically detects PD-L1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| PD-L1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| Human | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 29126 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| Hu124 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
| Human PD-L1/B7-H1 | |
| 100 μg | |
| Primary | |
| Detects Human PD-L1/B7-H1 in direct ELISAs. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Atezolizumab. This product is for research use only. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction