Learn More
R&D Systems™ Human PELI1/Pellino-1 (NP_065702.2) VersaClone cDNA
Product Code. 18703457
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18703457 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18703457 Supplier R&D Systems™ Supplier No. RDC3649
  • ONLINE EXCLUSIVE PRICE
    287.00€
    270.90€ / 10µg
    Save 16.10€
    5% Off
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
Specifications
Gene Pellino 1
Gene ID (Entrez) 57162
Gene Alias DKFZp686C18116, MGC50990, pellino (Drosophila) homolog 1, pellino homolog 1 (Drosophila), pellino-1, Pellino-related intracellular-signaling molecule, PRISM, protein pellino homolog 1
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg