Learn More
Human Phospholipid Scramblase 1/PLSCR1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB8137-SP
This item is not returnable.
View return policy
Description
Phospholipid Scramblase 1/PLSCR1 Monoclonal specifically detects Phospholipid Scramblase 1/PLSCR1 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Phospholipid Scramblase 1/PLSCR1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| O15162 | |
| PLSCR1 | |
| E. coli-derived recombinant human Phospholipid Scramblase 1/PLSCR1 Met1-Pro84 Accession # O15162 | |
| 25 μg | |
| Primary | |
| Detects human Phospholipid Scramblase 1/PLSCR1 in ELISA. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 875327 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Erythrocyte phospholipid scramblase, MmTRA1b, MMTRA1BCa(2+)-dependent phospholipid scramblase 1, phospholipid scramblase 1, PL scramblase 1, PLSCR1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5359 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |