missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PKC theta Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC4368T-100UG
This item is not returnable.
View return policy
Description
PKC theta Monoclonal specifically detects PKC theta in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| PKC theta | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q04759 | |
| PRKCQ | |
| E. coli-derived recombinant human PKC theta His580-Ser706 Accession # Q04759 | |
| RUO | |
| 5588 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 453416 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| EC 2.7.11, EC 2.7.11.13, MGC126514, MGC141919, nPKC-theta, PRKCQ, PRKCT, protein kinase C theta type, protein kinase C, theta | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human PKC theta . This antibody also detects recombinant human PKC theta , but does not detect any other recombinant human PKC family member. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction