missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pro BDNF Antibody, R&D Systems™

Mouse Monoclonal Antibody
201.00€ - 403.00€
Specifications
| Antigen | BDNF |
|---|---|
| Clone | 584431 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18141681
|
R&D Systems
MAB31752-SP |
25 μg |
201.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18122931
![]() |
R&D Systems
MAB31752-100 |
100 μg |
403.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BDNF Monoclonal specifically detects BDNF in Human samples. It is validated for ELISA.Specifications
| BDNF | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P23560 | |
| 627 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 584431 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| abrineurin, ANON2, brain-derived neurotrophic factor, BULN2, MGC34632, neurotrophin | |
| Chinese hamster ovary cell line CHO-derived recombinant human Pro-BDNF Met1-Arg247 Accession # P23560 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human Pro-BDNF in direct ELISAs and sandwich immunoassays. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title