missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Prolyl Oligopeptidase/PREP Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF4308
This item is not returnable.
View return policy
Description
Prolyl Oligopeptidase/PREP Polyclonal specifically detects Prolyl Oligopeptidase/PREP in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Prolyl Oligopeptidase/PREP | |
| Polyclonal | |
| Unconjugated | |
| P48147 | |
| PREP | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human PREP Leu2-Pro710 Accession # P48147 | |
| 100 μg | |
| Primary | |
| Detects human PREP in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant mouse PREP is observed. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| dJ355L5.1 (prolyl endopeptidase), EC 3.4.21.26, MGC16060, PE, PEP, Post-proline cleaving enzyme, PREP, prolyl endopeptidase, prolyl oligopeptidase, rPop | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5550 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction