missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Protein Disulfide Isomerase/P4HB Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF4236
This item is not returnable.
View return policy
Description
Protein Disulfide Isomerase/P4HB Polyclonal specifically detects Protein Disulfide Isomerase/P4HB in Human samples. It is validated for Western Blot, Simple Western, Immunoprecipitation, Immunocytochemistry.
Specifications
| Protein Disulfide Isomerase/P4HB | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cellular thyroid hormone-binding protein, collagen prolyl 4-hydroxylase beta, DSI, ERBA2L, GIT, glutathione-insulin transhydrogenase, P4Hbeta, p55, PDI, PDIA1, PDIA1procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase-associated 1), PDIEC 5.3.4.1, PHDB, PO4DB, PO4HB, procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide, PROHB, PROHBprocollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), betapolypeptide (protein disulfide isomerase; thyroid hormone binding protein p55), Prolyl 4-hydroxylase subunit beta, prolyl 4-hydroxylase, beta polypeptide, protein disulfide isomerase family A, member 1, protein disulfide isomerase/oxidoreductase, protein disulfide isomerase-associated 1, protein disulfide-isomerase, protocollagen hydroxylase, Thbp, thyroid hormone-binding protein p55 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5034 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 1-15 ug/mL | |
| P07237 | |
| P4HB | |
| Chinese hamster ovary cell line CHO-derived recombinant human P4HB Asp18-Lys505 Accession # P07237 | |
| 100 μg | |
| Primary | |
| Detects human P4HB in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction