missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Protocadherin-19 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB8626A-025
This item is not returnable.
View return policy
Description
Protocadherin-19 Monoclonal specifically detects Protocadherin-19 in Human samples. It is validated for Flow Cytometry.
Specifications
| Protocadherin-19 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| PCDH19 | |
| HEK293 human embryonic kidney cell line transfected with human Protocadherin-19 Met1-Ser678 Accession # Q8TAB3 | |
| 25 Tests | |
| Primary | |
| Detects human Protocadherin-19 in direct ELISA and flow cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 921614 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Q8TAB3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 57526 | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction