missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PTP rho/PTPRT Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3697
This item is not returnable.
View return policy
Description
PTP rho/PTPRT Polyclonal specifically detects PTP rho/PTPRT in Human samples. It is validated for Western Blot.
Specifications
| PTP rho/PTPRT | |
| Polyclonal | |
| Unconjugated | |
| O14522 | |
| PTPRT | |
| Mouse myeloma cell line NS0-derived recombinant human PTP rho/PTPRT Gln26-Lys725 Accession # O14522 | |
| 100 μg | |
| Primary | |
| Detects human PTP rho/PTPRT in Western blots. In Western blots, no cross-reactivity with recombinant human LAR, PTPRM, K, S, G or DEP1/CD148 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| EC 3.1.3, EC 3.1.3.48, KIAA0283RPTPrho, protein tyrosine phosphatase, receptor type, T, PTPRT, receptor protein tyrosine phosphatase, Receptor-type tyrosine-protein phosphatase rho, receptor-type tyrosine-protein phosphatase T, RPTP-rho, R-PTP-T | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 11122 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction