missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Respiratory Syncytial Virus A2 Fusion Protein Antibody (R338) [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89761JF525
This item is not returnable.
View return policy
Description
Human Respiratory Syncytial Virus A2 Fusion Protein Monoclonal antibody specifically detects Human Respiratory Syncytial Virus A2 Fusion Protein in Virus samples. It is validated for Western Blot,Sandwich ELISA Capture,Neutralization
Specifications
| Human Respiratory Syncytial Virus A2 Fusion Protein | |
| Monoclonal | |
| Janelia Fluor 525 | |
| HRSVgp08, RSV, RSV fusion protein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Respiratory Syncytial Virus (Uniprot#: P03420.1; Met1-Thr529) | |
| 0.2 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Sandwich ELISA Capture, Neutralization | |
| R338 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction