missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Human, Reverse Primer, Desalted

Product Code. 15839866 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
25 nmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15839866 25 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15839866 Supplier Applied Biosystems™ Supplier No. A15630

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Human, Reverse Primer, Desalted

Use our tool to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and dimers, target-specific, and suitable for universal PCR conditions, these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All primers are checked by mass spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Delivery Type Single Tube, Dried-down
For Use With (Equipment) Mass Spectrometry
Description Reverse PCR/SEQ Primer, 25 nmole, Solid, Desalted Purification, Hs00196555_CE Primer ID, 8nmole Minimal Spec Yield, Non-tailed Primer 5'-tail Option, Human Species, Delivered in a Single tube, Dried-down, Used for PCR and Cycle Sequencing Reactions
Product Type Reverse PCR or SEQ Primer
Selection Agent (Eukaryotic) Human
Purification Method Desalted
Primer Reverse PCR/SEQ
Quantity 25 nmol
Grade HPLC
For Use With (Application) PCR and Cycle Sequencing Reactions
Form Solid
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.