missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ROR1 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems FAB2000G-100UG
This item is not returnable.
View return policy
Description
ROR1 Polyclonal antibody specifically detects ROR1 in Human samples. It is validated for Flow Cytometry
Specifications
| ROR1 | |
| Polyclonal | |
| Flow Cytometry 5 uL/10^6 cells | |
| Q01973 | |
| Goat | |
| Antigen Affinity-purified | |
| RUO | |
| 4919 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG |
| Flow Cytometry | |
| Alexa Fluor 488 | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| EC 2.7.10.1, MGC99659, neurotrophic tyrosine kinase receptor-related 1, Neurotrophic tyrosine kinase, receptor-related 1, NTRKR1, NTRKR1dJ537F10.1, receptor tyrosine kinase-like orphan receptor 1, tyrosine-protein kinase transmembrane receptor ROR1 | |
| Mouse myeloma cell line NS0-derived recombinant human ROR1, Ala24-Glu403, Accession # Q01973 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction