missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human S100A6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7467-SP
This item is not returnable.
View return policy
Description
S100A6 Monoclonal specifically detects S100A6 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| S100A6 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P06703 | |
| S100A6 | |
| E. coli-derived recombinant human S100A6 Met1-Gly90 Accession # P06703 | |
| 25 μg | |
| Primary | |
| Detects human S100A6 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse S100A6 and no cross-reactivity with recombinant human (rh) S100A1, rhS100A4, or rhS100B is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 754102 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 2A9, CABP, CACY, CACY5B10, calcyclin, Growth factor-inducible protein 2A9, MLN 4, PRA, PRAS100 calcium binding protein A6 (calcyclin), Prolactin receptor-associated protein, protein S100-A6, S100 calcium binding protein A6, S100 calcium-binding protein A6, S100 calcium-binding protein A6 (calcyclin) | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6277 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction