Learn More
ERK5/BMK1 Antibody (OTI3H2), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-71243APC
Product Code. 18171352
  • 807.00€ / 0.10mL
Description
ERK5/BMK1 Monoclonal specifically detects ERK5/BMK1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ERK5/BMK1
Monoclonal
APC
PBS with 0.05% Sodium Azide
MAPK7
Full length recombinant protein of human ERK5/BMK1 (NP_002740) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Flow Cytometry
OTI3H2
Flow Cytometry
Big MAP kinase 1, BMK-1, BMK1 kinase, BMK1EC 2.7.11.24, EC 2.7.11, ERK-5, ERK5MAPK 7, Extracellular signal-regulated kinase 5, extracellular-signal-regulated kinase 5, MAP kinase 7, mitogen-activated protein kinase 7, PRKM7ERK4
Mouse
Affinity Purified
Cell Cycle and Replication, Phospho Specific, Signal Transduction
5598.0
Store at 4°C in the dark.