missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SHARPIN Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB8100
This item is not returnable.
View return policy
Description
SHARPIN Monoclonal specifically detects SHARPIN in Human samples. It is validated for Western Blot.
Specifications
| SHARPIN | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| Q9H0F6 | |
| SHARPIN | |
| E. coli-derived recombinant human SHARPIN Ala27-Ala182 Accession # Q9H0F6 | |
| 100 μg | |
| Primary | |
| Detects human SHARPIN in ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 881816 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp434N1923, hSIPL1, SHANK-associated RH domain interactor, Shank-associated RH domain-interacting protein, Shank-interacting protein-like 1, sharpin, SIPL1, SIPL1DKFZP434N1923 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 81858 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction